OriGene
Manufacturer Code:CF806723
Catalog # 50-166-7949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 180-482 of human DUSP10 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dual specificity phosphatase MKP-5; Dual specificity protein phosphatase 10; MAP kinase phosphatase 5; Mitogen-activated protein kinase phosphatase 5; MKP-5 MKP5; serine/threonine specific protein phosphatase
Gene Aliases: DUSP10; MKP-5; MKP5
UniProt ID: (Human) Q9Y6W6
Entrez Gene ID: (Human) 11221
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.