Novus Biologicals
Manufacturer Code:NB100888
Catalog # NB100888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CRILTPKLMGVETVV corresponding to C-Terminus according to NP_009138.1 NP_653329.1 NP_653330.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity phosphatase 10 dual specificity phosphatase MKP-5 EC 3.1.3.16 EC 3.1.3.48 MAP kinase phosphatase 5 Mitogen-activated protein kinase phosphatase 5 MKP-5dual specificity protein phosphatase 10 MKP5serine/threonine specific protein phosphatase; dual specificity phosphatase MKP-5; Dual specificity protein phosphatase 10; MAP kinase phosphatase 5; Mitogen-activated protein kinase phosphatase 5; serine/threonine specific protein phosphatase
Gene Aliases: DUSP10; MKP-5; MKP5
UniProt ID: (Human) Q9Y6W6
Entrez Gene ID: (Human) 11221
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.