Invitrogen
Manufacturer Code:PA571497
Catalog # PIPA571497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 513-542 amino acids from the Central region of human DUOX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual oxidase 2; Dual oxidase 2 1111- Large NOX 2 Long NOX 2 NADH/NADPH thyroid oxidase p138-tox NADPH oxidase/peroxidase DUOX2 NADPH thyroid oxidase 2 Thyroid oxidase 2 p138 thyroid oxidase DUOX2 LNOX2 THOX2; dual oxidase-like domains 2; flavoprotein NADPH oxidase; Large NOX 2; Long NOX 2; NADH/NADPH thyroid oxidase p138-tox; NADPH oxidase/peroxidase DUOX2; NADPH thyroid oxidase 2; nicotinamide adenine dinucleotide phosphate oxidase; p138 thyroid oxidase; Thyroid oxidase 2
Gene Aliases: DUOX2; LNOX2; NOXEF2; P138-TOX; TDH6; THOX2
UniProt ID: (Human) Q9NRD8
Entrez Gene ID: (Human) 50506
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.