Novus Biologicals
Manufacturer Code:NB11061576AF700
Catalog # NB11061576Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-Prot# Q9NRD8] |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual oxidase 2; dual oxidase 2 dual oxidase-like domains 2 EC 1.11.1.- EC 1.6.3.1 flavoprotein NADPH oxidase Large NOX 2 LNOX2P138(TOX) Long NOX 2 NADH/NADPH thyroid oxidase p138-tox NADPH oxidase/peroxidase DUOX2 NADPH thyroid oxidase 2 nicotinamide adenine dinucleotide phosphate oxidase NOXEF2 p138 thyroid oxidase P138-TOX THOX2TDH6 Thyroid oxidase 2; dual oxidase-like domains 2; flavoprotein NADPH oxidase; Large NOX 2; Long NOX 2; NADH/NADPH thyroid oxidase p138-tox; NADPH oxidase/peroxidase DUOX2; NADPH thyroid oxidase 2; nicotinamide adenine dinucleotide phosphate oxidase; p138 thyroid oxidase; Thyroid oxidase 2
Gene Aliases: DUOX2; LNOX2; NOXEF2; P138-TOX; TDH6; THOX2
UniProt ID: (Human) Q9NRD8
Entrez Gene ID: (Human) 50506
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.