Invitrogen
Manufacturer Code:PA571444
Catalog # PIPA571444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human DUOX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual oxidase 1; flavoprotein NADPH oxidase; Large NOX 1; LNOX1 NOXEF1 THOX1 NADPH thyroid oxidase 1 flavoprotein NADPH oxidase large NOX 1 long NOX 1 nicotinamide adenine dinucleotide phosphate oxidase thyroid oxidase 1; Long NOX 1; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; Thyroid oxidase 1
Gene Aliases: DUOX; DUOX1; LNOX1; NOXEF1; THOX1
UniProt ID: (Human) Q9NRD9
Entrez Gene ID: (Human) 53905
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.