Novus Biologicals
Manufacturer Code:NBP216232
Catalog # NBP216232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DUOX1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual oxidase 1; dual oxidase 1 DUOX EC 1.11.1.- EC 1.6.3.1 Large NOX 1 LNOX1flavoprotein NADPH oxidase Long NOX 1 MGC138840 NADPH thyroid oxidase 1 nicotinamide adenine dinucleotide phosphate oxidase NOXEF1 THOX1MGC138841 Thyroid oxidase 1; flavoprotein NADPH oxidase; Large NOX 1; Long NOX 1; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; Thyroid oxidase 1
Gene Aliases: DUOX; DUOX1; LNOX1; NOXEF1; THOX1
UniProt ID: (Human) Q9NRD9
Entrez Gene ID: (Human) 53905
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.