Abnova
Manufacturer Code:H00053905A01
Catalog # 89-106-951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 941 a.a. to 1028 a.a) partial recombinant protein with GST tag.,DUOX1 (NP_059130 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant DUOX1.
For Research Use Only
Protein Aliases: Dual oxidase 1; flavoprotein NADPH oxidase; Large NOX 1; LNOX1; Long NOX 1; MGC138840; MGC138841; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF1; THOX1; Thyroid oxidase 1
Gene Aliases: DUOX; DUOX1; LNOX1; NOXEF1; THOX1
UniProt ID: (Human) Q9NRD9
Entrez Gene ID: (Human) 53905
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.