Novus Biologicals
Manufacturer Code:NBP201184
Catalog # NBP201184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G8 |
Immunogen | Full length human recombinant protein of human DTYMK(NP_036277) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDC8TMPKTYMKPP3731 deoxythymidylate kinase (thymidylate kinase) dTMP kinase EC 2.7.4.9 FLJ44192 MGC198617 thymidylate kinase; deoxythymidylate kinase (thymidylate kinase); dTMP kinase; Thymidylate kinase
Gene Aliases: CDC8; DTYMK; PP3731; TMPK; TYMK
UniProt ID: (Human) P23919
Entrez Gene ID: (Human) 1841
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.