OriGene
Manufacturer Code:CF503497
Catalog # 50-166-2177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D2 |
Immunogen | Full length human recombit protein of human DTYMK produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDC8 PP3731 TMPK TYMK; deoxythymidylate kinase (thymidylate kinase); dTMP kinase; Thymidylate kinase
Gene Aliases: CDC8; DTYMK; PP3731; TMPK; TYMK
UniProt ID: (Human) P23919
Entrez Gene ID: (Human) 1841
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.