Proteintech
Manufacturer Code:183501AP150UL
Catalog # 50-173-2129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DTX1 Fusion Protein Ag13172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deltex 1; deltex 1, E3 ubiquitin ligase; deltex homolog 1; deltex homolog 1 (Drosophila) Deltex1 DTX1 hDTX1 hDx 1 Protein deltex 1; Deltex1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; FXI-T1; hDTX1; mDTX1; Protein deltex-1; RING-type E3 ubiquitin transferase DTX1
Gene Aliases: DTX1; Fxit1; hDx-1; mKIAA4160
UniProt ID: (Human) Q86Y01, (Mouse) Q61010
Entrez Gene ID: (Human) 1840, (Rat) 687424, (Mouse) 14357
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.