Invitrogen
Manufacturer Code:PA582998
Catalog # PIPA582998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DTNBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; Biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; BLOC1S8 DBND Dysbindin HPS7 My031; Dysbindin; Dysbindin-1; Dystrobrevin-binding protein 1; Hermansky-Pudlak syndrome 7 protein; HPS7 protein
Gene Aliases: BLOC1S8; DBND; DTNBP1; HPS7; My031; SDY
UniProt ID: (Human) Q96EV8
Entrez Gene ID: (Human) 84062
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.