Novus Biologicals
Manufacturer Code:NBP23532520UL
Catalog # NBP23532520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Human DTD1. Located within the following region: APGTATSDPKQLSKLEKQQQRKEKTRAKGPSESSKERNTPRKEDRSASSG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA379J5.3 bA555E18.1 C20orf88HARS2 chromosome 20 open reading frame 88 DNA-unwinding element-binding protein D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae) D-tyrosyl-tRNA(Tyr) deacylase 1 DUEB EC 3.1 Histidyl-tRNA synthase-related histidyl-tRNA synthetase 2 MGC119131 MGC41905 pqn-68; D-aminoacyl-tRNA deacylase 1; D-tyrosyl-tRNA deacylase 1 homolog; DNA-unwinding element-binding protein B; DTD; DUE-B; Gly-tRNA(Ala) deacylase; Histidyl-tRNA synthase-related; histidyl-tRNA synthetase 2
Gene Aliases: C20orf88; DTD1; DUE-B; DUEB; HARS2; pqn-68
UniProt ID: (Human) Q8TEA8
Entrez Gene ID: (Human) 92675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.