OriGene
Manufacturer Code:TA890114
Catalog # 50-168-4800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human DSP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 250/210 kDa paraneoplastic pemphigus antigen; DCWHKTA DP; Desmoplakin; DP
Gene Aliases: DCWHKTA; DP; DSP
UniProt ID: (Human) P15924
Entrez Gene ID: (Human) 1832
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.