Proteintech
Manufacturer Code:218801AP150UL
Catalog # 50-173-3622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DSG2 Fusion Protein Ag16393 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARVC10 ARVD10 Cadherin family member 5 CDHF5 desmoglein 2 DSG2 HDGC; Cadherin family member 5; desmoglein 2; Desmoglein-2; HDGC
Gene Aliases: AA408168; CDHF5; D18Ertd293e; DSG2; HDGC
UniProt ID: (Human) Q14126, (Mouse) Q8K069
Entrez Gene ID: (Human) 1829, (Mouse) 13511, (Rat) 307562
Molecular Function:
cadherin
cell adhesion molecule
cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.