Proteintech
Manufacturer Code:600411IG150UL
Catalog # 50-173-6200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C9F7 |
Immunogen | DSE Fusion Protein Ag0695 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin-glucuronate 5-epimerase; dermatan sulfate epimerase DS epimerase DSE DSEPI SART 2 SART2; Dermatan-sulfate epimerase; DS epimerase; SART-2; squamous cell carcinoma antigen recognized by T cells 2; Squamous cell carcinoma antigen recognized by T-cells 2
Gene Aliases: 6030499O08; AI480506; B130024B19Rik; DS-epi1; DSE; DSEP; DSEPI; EDSMC2; SART-2; SART2
UniProt ID: (Human) Q9UL01, (Mouse) Q8BLI4
Entrez Gene ID: (Human) 29940, (Mouse) 212898
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.