Invitrogen
Manufacturer Code:MA538045
Catalog # PIMA538045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1918 |
Immunogen | A synthesized peptide derived from human CRMP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 6330417M19Rik AI450666 C-terminal region Dlp1 DNM1L Dnm1p/Vps1p-like protein Dnml1 Dnmlp1 Drp1 DVLP DYMPLE dynamin 1 like dynamin 1-like dynamin family member proline-rich carboxyl-terminal domain less Dynamin-1-like protein Dynamin-like protein dynamin-like protein 1 dynamin-like protein 4 Dynamin-like protein IV Dynamin-related protein 1 DYNIV 11 DYNIV-11 EC 3.6.5.5 EMPF FLJ41912 HDYNIV N-terminal region python VPS1; C-terminal region; Dnm1p/Vps1p-like protein; DVLP; Dymple; dynamin 1-like; Dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-like protein; dynamin-like protein 1; Dynamin-like protein 4; Dynamin-like protein IV; Dynamin-related protein 1; HdynIV; N-terminal region
Gene Aliases: DLP1; DNM1L; Dnml1; DRP1; DVLP; DYMPLE; EMPF; HDYNIV
UniProt ID: (Human) O00429, (Rat) O35303
Entrez Gene ID: (Human) 10059, (Rat) 114114
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.