Invitrogen
Manufacturer Code:PA520175
Catalog # PIPA520175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide from near the carboxy terminus of human CRMP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collapsin response mediator protein 1; CRMP-1; CRMP-1 DPYSL1 DRP-1 DRP1 ULIP-3 dihydropyrimidinase related protein-1 dihydropyrimidinase-like 1 dihydropyrimidinase-related protein 1 unc-33-like phosphoprotein 3; dihydropyrimidinase-like 1; Dihydropyrimidinase-related protein 1; DRP-1; Inactive dihydropyrimidinase; ULIP-3; Unc-33-like phosphoprotein 3
Gene Aliases: CRMP-1; CRMP1; DPYSL1; DRP-1; DRP1; ULIP-3; ULIP3
UniProt ID: (Human) Q14194, (Rat) Q62950, (Mouse) P97427
Entrez Gene ID: (Human) 1400, (Rat) 25415, (Mouse) 12933
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.