Novus Biologicals
Manufacturer Code:NB11055237
Catalog # NB11055237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an N-terminal region within residues 1-100 of the human DRP1 protein. [Swiss-Prot# O00429] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DLP1 DRP1Dnm1p/Vps1p-like protein DVLPEC 3.6.5.5 Dymple DYMPLEDYNIV-11 dynamin 1-like Dynamin family member proline-rich carboxyl-terminal domain less dynamin-1-like protein Dynamin-like protein Dynamin-like protein 4 Dynamin-like protein IV Dynamin-related protein 1 FLJ41912 HDYNIV VPS1; Dnm1p/Vps1p-like protein; DVLP; Dymple; dynamin 1-like; Dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-like protein; Dynamin-like protein 4; Dynamin-like protein IV; Dynamin-related protein 1; HdynIV
Gene Aliases: DLP1; DNM1L; DRP1; DVLP; DYMPLE; EMPF; HDYNIV
UniProt ID: (Human) O00429
Entrez Gene ID: (Human) 10059
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.