Invitrogen
Manufacturer Code:PA569487
Catalog # PIPA569487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human DRGX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dorsal root ganglia homeobox protein; Dorsal root ganglia homeobox protein Paired-related homeobox protein-like 1 DRGX PRRXL1; Dorsal root ganglion 11; Homeobox protein DRG11; paired homeodomain protein DRG11; paired related homeobox protein-like 1; paired related homeobox protein-like 1b; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; Paired-related homeobox protein-like 1
Gene Aliases: DRG11; DRGX; PRRXL1
UniProt ID: (Human) A6NNA5, (Rat) Q62798, (Mouse) Q8BYH0
Entrez Gene ID: (Human) 644168, (Pig) 100154590, (Horse) 100051674, (Rabbit) 100345527, (Dog) 607056, (Rat) 252880, (Guinea pig) 100719763, (Mouse) 107751
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.