Abnova
Manufacturer Code:PAB25731
Catalog # 89-127-355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at C-terminus of human DRGX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against synthetic peptide of DRGX.
For Research Use Only
Protein Aliases: Dorsal root ganglia homeobox protein; DRG11; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; Paired-related homeobox protein-like 1; PRRXL1
Gene Aliases: DRG11; DRGX; PRRXL1
UniProt ID: (Human) A6NNA5
Entrez Gene ID: (Human) 644168
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.