Novus Biologicals
Manufacturer Code:NBP271419G
Catalog # NBP271419G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A2 |
Immunogen | Full length recombinant protein of human DREF (NP_004720) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Ac-like transposable element; Ac-like transposable element ALTEDNA replication-related element binding factor hDREF KIAA0785dREF homolog Putative Ac-like transposable element putative Ac-like transposon TRAMPDREFBED-type zinc finger domain-containing protein 1 zinc finger BED domain-containing protein 1 zinc finger BED-type containing 1; BED-type zinc finger domain-containing protein 1; DNA replication-related element binding factor; DNA replication-related element-binding factor; dREF homolog; E3 SUMO-protein ligase ZBED1; Putative Ac-like transposable element; Zinc finger BED domain-containing protein 1; zinc finger, BED-type containing 1
Gene Aliases: ALTE; DREF; hDREF; KIAA0785; TRAMP; ZBED1
UniProt ID: (Human) O96006
Entrez Gene ID: (Human) 9189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.