Invitrogen
Manufacturer Code:PA572237
Catalog # PIPA572237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 421-456 amino acids from the C-terminal region of human DRD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(1B) dopamine receptor; D(5) dopamine receptor; D1beta dopamine receptor; Dopamine D5 receptor; dopamine receptor D1B; DRD5 D(5) dopamine receptor D1beta dopamine receptor D5 dopamine receptor Dopamine 5 receptor Dopamine receptor D5 DRD1B D(1B) dopamine receptor D1b dopamine receptor D1b receptor DBDR Dopamine receptor 5 DRD1L2 Hd5dr Dopamine d1b receptor Dopamine D5 receptor Dopamine receptor D1B
Gene Aliases: DBDR; DRD1B; DRD1L2; DRD5
UniProt ID: (Human) P21918
Entrez Gene ID: (Human) 1816
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.