Invitrogen
Manufacturer Code:PA532378
Catalog # PIPA532378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to aa 459-477 of human D5DR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(1B) dopamine receptor; D(5) dopamine receptor; D1beta dopamine receptor; DBDR DRD1B DRD1L2 D1B dopamine receptor D1beta dopamine receptor d5 dopamine receptor dopamine D5 receptor dopamine receptor D1B; Dopamine D5 receptor; dopamine receptor D1B
Gene Aliases: DBDR; DRD1B; DRD1L2; DRD5
UniProt ID: (Human) P21918
Entrez Gene ID: (Human) 1816
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.