Abnova
Manufacturer Code:PAB26234
Catalog # 89-127-875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human DRD5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DRD5.
For Research Use Only
Protein Aliases: D(1B) dopamine receptor; D(5) dopamine receptor; D1beta dopamine receptor; DBDR; Dopamine D5 receptor; dopamine receptor D1B; DRD1B; DRD1L2; MGC10601
Gene Aliases: DBDR; DRD1B; DRD1L2; DRD5
UniProt ID: (Human) P21918
Entrez Gene ID: (Human) 1816
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.