Proteintech
Manufacturer Code:203101AP150UL
Catalog # 50-173-2821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DRD5 Fusion Protein Ag14145 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(1B) dopamine receptor; D(1B) dopamine receptor D(5) dopamine receptor D1beta dopamine receptor DBDR Dopamine D5 receptor dopamine receptor D5 DRD1B DRD1L2 DRD5; D(5) dopamine receptor; D1beta dopamine receptor; Dopamine D5 receptor; dopamine receptor 5; dopamine receptor D1B
Gene Aliases: D1B; D5R; DBDR; Drd-5; DRD1B; DRD1L2; DRD5; Gpcr1
UniProt ID: (Human) P21918, (Mouse) Q8BLD9, (Rat) P25115
Entrez Gene ID: (Human) 1816, (Mouse) 13492, (Rat) 25195
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.