Invitrogen
Manufacturer Code:PA577390
Catalog # PIPA577390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CGAENSTGVNRARPH corresponding to amino acid residues 15-29 of rat D3 Dopamine receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(3) dopamine receptor; D3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopaminergic receptor D3; DRD3
Gene Aliases: D3R; Drd3
UniProt ID: (Rat) P19020, (Mouse) P30728
Entrez Gene ID: (Rat) 29238, (Mouse) 13490
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.