Invitrogen
Manufacturer Code:PA579170
Catalog # PIPA579170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Dopamine Receptor D3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(3) dopamine receptor; D(3) dopamine receptor Dopamine D3 receptor DRD3; D3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopaminergic receptor D3; essential tremor 1
Gene Aliases: D3DR; D3R; DRD3; ETM1; FET1
UniProt ID: (Human) P35462, (Rat) P19020, (Mouse) P30728
Entrez Gene ID: (Human) 1814, (Rat) 29238, (Mouse) 13490
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.