Invitrogen
Manufacturer Code:PA533482
Catalog # PIPA533482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human DRD3 / Dopamine Receptor D3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(3) dopamine receptor; Dopamine D3 receptor; DRD3 D3 dopamine receptor D3DR Dopamine 3 receptor Dopamine receptor D3 D3nf Essential tremor 1 ETM1 FET1 D(3) dopamine receptor Dopamine D3 receptor; essential tremor 1
Gene Aliases: D3DR; DRD3; ETM1; FET1
UniProt ID: (Human) P35462
Entrez Gene ID: (Human) 1814
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.