Invitrogen
Manufacturer Code:PA5115143
Catalog # PIPA5115143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DRD3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(3) dopamine receptor; D(3) dopamine receptor D3 receptor D3DR D3nf D3R Dopamine 3 receptor Dopamine D3 receptor dopamine receptor 3 dopamine receptor D3 dopaminergic receptor D3 DRD3 essential tremor 1 ETM1 FET1; D3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopaminergic receptor D3; essential tremor 1
Gene Aliases: D3DR; D3R; DRD3; ETM1; FET1
UniProt ID: (Human) P35462, (Rat) P19020, (Mouse) P30728
Entrez Gene ID: (Human) 1814, (Rat) 29238, (Mouse) 13490
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.