Invitrogen
Manufacturer Code:PA577389
Catalog # PIPA577389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DDLERQNWSRPFNGSE corresponding to amino acid residues 11-26 of rat D2 dopamine receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(2) dopamine receptor; D2 receptor; Dopamine D2 receptor; dopamine receptor 2; DRD2
Gene Aliases: D2R; Drd-2; Drd2
UniProt ID: (Mouse) P61168, (Rat) P61169
Entrez Gene ID: (Mouse) 13489, (Rat) 24318
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.