Invitrogen
Manufacturer Code:MA537607
Catalog # PIMA537607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 629CT62.2.2 |
Immunogen | KLH conjugated synthetic peptide between 307-336 amino acids from the C-terminal region of human DRD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D(2) dopamine receptor; D(2) dopamine receptor D2 receptor D2DR D2R Dopamine 2 receptor Dopamine D2 receptor dopamine receptor 2 dopamine receptor D2 dopamine receptor D2 isoform Drd2 Drd-2 seven transmembrane helix receptor; D2 receptor; Dopamine D2 receptor; dopamine receptor 2; dopamine receptor D2 isoform; seven transmembrane helix receptor
Gene Aliases: D2DR; D2R; Drd-2; DRD2
UniProt ID: (Human) P14416, (Mouse) P61168, (Rat) P61169
Entrez Gene ID: (Human) 1813, (Mouse) 13489, (Rat) 24318
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.