Invitrogen
Manufacturer Code:PA527172
Catalog # PIPA527172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 347 and 441 of Human Dopamine Receptor D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(1A) dopamine receptor; D1 receptor; D1A dopamine receptor; DADR DRD1A D(1A) dopamine receptor dopamine D1 receptor; Dopamine D1 receptor; dopamine receptor D1A; G protein-coupled receptor DRD1
Gene Aliases: C030036C15Rik; DADR; Drd-1; DRD1; DRD1A; Gpcr15
UniProt ID: (Human) P21728, (Mouse) Q61616
Entrez Gene ID: (Human) 1812, (Mouse) 13488
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.