Invitrogen
Manufacturer Code:720276
Catalog # PI720276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to Human DRD1 (aa 164-179) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(1A) dopamine receptor; D1 receptor; D1A dopamine receptor; Dopamine D1 receptor; Dopamine Receptor D1 D(1A) dopamine receptor; dopamine receptor D1A; Dopamine-1A receptor; G protein-coupled receptor DRD1
Gene Aliases: C030036C15Rik; D1a; DADR; Drd-1; DRD1; DRD1A; Gpcr15
UniProt ID: (Human) P21728, (Mouse) Q61616
Entrez Gene ID: (Human) 1812, (Mouse) 13488, (Rat) 24316
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.