R&D Systems
Manufacturer Code:MAB6640SP
Catalog # MAB6640SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 685228 |
Immunogen | <em>E. coli</em>-derived recombinant human DRAK2 Ala87-Glu285 (predicted) Accession # O94768 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DAP kinase-related apoptosis-inducing protein kinase 2; DAP kinase-related apoptosis-inducing protein kinase 2 DRAK2death-associated protein kinase-related 2 EC 2.7.11 EC 2.7.11.1 serine/threonine kinase 17b serine/threonine kinase 17b (apoptosis-inducing) serine/threonine-protein kinase 17B STK17B; death-associated protein kinase-related 2; Serine/threonine-protein kinase 17B
Gene Aliases: DRAK2; STK17B
UniProt ID: (Human) O94768
Entrez Gene ID: (Human) 9262
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.