Invitrogen
Manufacturer Code:PA577273
Catalog # PIPA577273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 261-310 of Human DRAK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAP kinase related apoptosis inducing protein DAP kinase related apoptosis inducing protein kinase 2 DAP kinase-related apoptosis-inducing protein kinase 2 Death associated protein kinase related 2 DRAK 2 DRAK2 Serine/threonine kinase 17b Serine/threonine kinase 17b apoptosis inducing Serine/threonine protein kinase 17B Serine/threonine-protein kinase 17B ST17B STK 17B Stk17b; DAP kinase-related apoptosis-inducing protein kinase 2; death-associated protein kinase-related 2; serine/threonine kinase 17b (apoptosis-inducing); Serine/threonine-protein kinase 17B
Gene Aliases: 3110009A03Rik; AI120141; DRAK2; STK17B
UniProt ID: (Human) O94768, (Mouse) Q8BG48, (Rat) Q91XS8
Entrez Gene ID: (Human) 9262, (Mouse) 98267, (Rat) 170904
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.