Novus Biologicals
Manufacturer Code:NBP184980
Catalog # NBP184980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ICLSHSWLQQWDFENLFHPEETSSSSQTQDHSVRSSEDKTSKSSCNGTCGDREDKENIPEDSSMVSKRFRFDDSLPNPHELVSDLL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAP kinase-related apoptosis-inducing protein kinase 2; DAP kinase-related apoptosis-inducing protein kinase 2 DRAK2death-associated protein kinase-related 2 EC 2.7.11 EC 2.7.11.1 serine/threonine kinase 17b serine/threonine kinase 17b (apoptosis-inducing) serine/threonine-protein kinase 17B; death-associated protein kinase-related 2; Serine/threonine-protein kinase 17B
Gene Aliases: DRAK2; STK17B
UniProt ID: (Human) O94768
Entrez Gene ID: (Human) 9262
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.