Novus Biologicals
Manufacturer Code:NB500345UV
Catalog # NB500345UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DR-6-04-EC |
Immunogen | A fusion protein representing amino acids 42-335 (extracellular part) of human DR6 linked to the Fc portion of human IgG1 was used as an immunogen. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BM-018 CD358 antigen Death receptor 6 DR6TNFR-related death receptor 6 MGC31965 tumor necrosis factor receptor superfamily member 21 tumor necrosis factor receptor superfamily member 21; CD358; Death receptor 6; TNFR-related death receptor 6; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21
Gene Aliases: BM-018; CD358; DR6; TNFRSF21; UNQ437/PRO868
UniProt ID: (Human) O75509
Entrez Gene ID: (Human) 27242
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.