Proteintech
Manufacturer Code:667541IG150UL
Catalog # 50-173-7161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C2D10 |
Immunogen | DR6 Fusion Protein Ag14327 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BM 018 CD358 Death receptor 6 DR6 TNFRSF21; CD358; Death receptor 6; TNFR-related death receptor 6; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21
Gene Aliases: BM-018; CD358; DR6; TNFRSF21; UNQ437/PRO868
UniProt ID: (Human) O75509
Entrez Gene ID: (Human) 27242
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.