Invitrogen
Manufacturer Code:MA531265
Catalog # PIMA531265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | Recombit Human TNFRSF21/DR6 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD358; Death receptor 6; Death receptor-6; TNFR-related death receptor 6; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21
Gene Aliases: BM-018; CD358; DR6; TNFRSF21; UNQ437/PRO868
UniProt ID: (Human) O75509
Entrez Gene ID: (Human) 27242
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.