Invitrogen
Manufacturer Code:PA563106
Catalog # PIPA563106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Down-regulator of transcription 1; Down-regulator of transcription 1 TBP-binding (negative cofactor 2) NC2 NC2-BETA; down-regulator of transcription 1, TBP-binding (negative cofactor 2); NC2-beta; negative cofactor 2; negative cofactor 2 beta; Negative cofactor 2-beta; Protein Dr1; TATA-binding protein-associated phosphoprotein
Gene Aliases: DR1; NC2; NC2-BETA; NC2B
UniProt ID: (Human) Q01658
Entrez Gene ID: (Human) 1810
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.