Invitrogen
Manufacturer Code:PA541427
Catalog # PIPA541427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Down-regulator of transcription 1; Down-regulator of transcription 1 TBP-binding (negative cofactor 2) NC2 NC2-BETA; down-regulator of transcription 1, TBP-binding (negative cofactor 2); NC2-beta; negative cofactor 2; negative cofactor 2 beta; Negative cofactor 2-beta; Protein Dr1; TATA-binding protein-associated phosphoprotein
Gene Aliases: 1700121L09Rik; DR1; Dr1l; NC2; NC2-BETA; NC2B; NC2beta; wu:fj15f04; zgc:152914
UniProt ID: (Human) Q01658, (Mouse) Q91WV0, (Rat) Q5XI68
Entrez Gene ID: (Bovine) 613828, (Horse) 100059248, (Human) 1810, (Dog) 479943, (Rabbit) 100352936, (Mouse) 13486, (Guinea pig) 100724062, (Rat) 289881, (Zebrafish) 768171
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.