OriGene
Manufacturer Code:CF811601
Catalog # 50-167-0741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 13-490 of human DPYSL2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP-2; CRMP-2 CRMP2 DHPRP2 DRP-2 DRP2 N2A3 ULIP-2 ULIP2; dihydropyrimidinase-like 2; Dihydropyrimidinase-related protein 2; DRP-2; N2A3; TOAD-64; Turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP-2; Unc-33-like phosphoprotein 2
Gene Aliases: AI851130; CRMP-2; CRMP2; DHPRP2; DPYSL2; DRP-2; DRP2; Musunc33; N2A3; TOAD-64; ULIP-2; ULIP2
UniProt ID: (Human) Q16555, (Rat) P47942, (Mouse) O08553
Entrez Gene ID: (Human) 1808, (Rat) 25416, (Mouse) 12934
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.