Abnova
Manufacturer Code:MAB12580
Catalog # 89-934-052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 33CT11.40.3 |
Immunogen | A synthetic peptide corresponding to amino acids 463-507 of human DPYSL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against human DPYSL2.
For Research Use Only
Protein Aliases: Collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP-2; CRMP2; DHPRP2; dihydropyrimidinase-like 2; Dihydropyrimidinase-related protein 2; DRP-2; DRP2; N2A3; ULIP-2; Unc-33-like phosphoprotein 2
Gene Aliases: CRMP-2; CRMP2; DHPRP2; DPYSL2; DRP-2; DRP2; N2A3; ULIP-2; ULIP2
UniProt ID: (Human) Q16555
Entrez Gene ID: (Human) 1808
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.