Novus Biologicals
Manufacturer Code:NBP191292AF532
Catalog # NP191292A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8D12 |
Immunogen | The immunogen for this antibody is DPYS. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DHP; DHPasehydantoinase DHPHydantoinase dihydropyrimidinase Dihydropyrimidine amidohydrolase EC 3.5.2.2; Dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Hydantoinase
Gene Aliases: DHP; DHPase; DPYS
UniProt ID: (Human) Q14117
Entrez Gene ID: (Human) 1807
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.