Proteintech
Manufacturer Code:132371AP150UL
Catalog # 50-172-8595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DPYS Fusion Protein Ag3972 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHP; DHP DHPase Dihydropyrimidinase DPYS Hydantoinase; DHPase; Dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Hydantoinase
Gene Aliases: 1200017I10Rik; 1300004I01Rik; DHP; DHPase; DPYS
UniProt ID: (Human) Q14117, (Rat) Q63150, (Mouse) Q9EQF5
Entrez Gene ID: (Human) 1807, (Rat) 65135, (Mouse) 64705
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.