Invitrogen
Manufacturer Code:PA579168
Catalog # PIPA579168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human DPYD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHPDHase; Dihydropyrimidine dehydrogenase &lsqbNADP(+)] DHPDHase DPD Dihydrothymine dehydrogenase Dihydrouracil dehydrogenase DPYD; Dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; Dihydrouracil dehydrogenase; DPD
Gene Aliases: AI315208; DHP; DHPDHASE; DPD; DPYD; E330028L06Rik
UniProt ID: (Human) Q12882, (Rat) O89000, (Mouse) Q8CHR6
Entrez Gene ID: (Human) 1806, (Rat) 81656, (Mouse) 99586
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.