Invitrogen
Manufacturer Code:PA552545
Catalog # PIPA552545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DPY19L3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dpy-19-like 3; Dpy-19-like protein 3; Probable C-mannosyltransferase DPY19L3; Probable C-mannosyltransferase DPY19L3 241- Dpy-19-like protein 3 Protein dpy-19 homolog 3 DPY19L3; Protein dpy-19 homolog 3
Gene Aliases: DPY19L3
UniProt ID: (Human) Q6ZPD9
Entrez Gene ID: (Human) 147991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.