Novus Biologicals
Manufacturer Code:NBP2829190.1ML
Catalog # NB030848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human DPY19L3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686J17135 dpy-19-like 3 (C. elegans) dpy-19-like protein 3 MGC35440 protein dpy-19 homolog 3; dpy-19-like 3; Dpy-19-like protein 3; Probable C-mannosyltransferase DPY19L3; Protein dpy-19 homolog 3
Gene Aliases: DPY19L3
UniProt ID: (Human) Q6ZPD9
Entrez Gene ID: (Human) 147991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.