Invitrogen
Manufacturer Code:PA544405
Catalog # PIPA544405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DPY19L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dpy-19-like 2; dpy-19-like 2 (C. elegans); Dpy-19-like protein 2; Probable C-mannosyltransferase DPY19L2; probable C-mannosyltransferase DPY19L2-like; Protein dpy-19 homolog 2; spermatogenesis associated 34
Gene Aliases: 4932443J21Rik; DPY19L2; Dyp19l2; Gm18376; RGD1564311; SPATA34; SPGF9; UNQ3127/PRO10284
UniProt ID: (Human) Q6NUT2, (Mouse) P0CW70
Entrez Gene ID: (Human) 283417, (Dog) 475282, (Bovine) 524676, (Rat) 300461, (Mouse) 320752
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.