Invitrogen
Manufacturer Code:PA569240
Catalog # PIPA569240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse DPY19L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dpy-19 like 1; dpy-19-like 1; DPY-19-like protein 1; DPY-19-like protein 1 protein dpy-19 homolog 1; Probable C-mannosyltransferase DPY19L1; Protein dpy-19 homolog 1
Gene Aliases: 1100001I19Rik; 6030402K12; AI225861; AW558481; DPY19L1; GA0500; KIAA0877; RGD1305822
UniProt ID: (Human) Q2PZI1, (Mouse) A6X919
Entrez Gene ID: (Rabbit) 100340917, (Human) 23333, (Dog) 482395, (Horse) 100070313, (Guinea pig) 100727671, (Mouse) 244745, (Rat) 315496
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.